Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
25 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 650,00 Kč |
O této položce
5 650,00 Kč
material
colorless , colorless wells, polycarbonate
feature
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128508
technique(s)
PCR: suitable
size
384 wells
working volume
40 μL
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| material colorless , polycarbonate , colorless wells | material conical bottom green wells, green plate, polypropylene | material conical bottom yellow wells, polypropylene , yellow plate | material polypropylene , red (wells colorless), red wells |
| manufacturer/tradename Eppendorf® 0030128508 | manufacturer/tradename Eppendorf® 0030128524 | manufacturer/tradename Eppendorf® 0030128516 | manufacturer/tradename Eppendorf® 0030128540 |
| working volume 40 μL | working volume - | working volume - | working volume - |
| size 384 wells | size 384 wells | size 384 wells | size 384 wells |
| packaging pkg of 25 ea | packaging pkg of 25 ea | packaging pkg of 25 ea | packaging pkg of 25 ea |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable |
Still not finding the right product?
Explore all of our products under Eppendorf® Twin tec PCR plates, 384 wells
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?