EP0030128540
Eppendorf® Twin tec PCR plates, 384 wells
red (wells colorless), pkg of 25 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
polypropylene
red (wells colorless)
red wells
red (wells colorless)
red wells
size:
384 well
feature:
skirt
Doporučené produkty
material
polypropylene
red (wells colorless)
red wells
feature
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128540
technique(s)
PCR: suitable
size
384 well
well maximum volume
40 μm
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)