Key Documents
EP0030128516
Eppendorf® Twin tec PCR plates, 384 wells
yellow plate, conical bottom yellow wells, pkg of 25 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material
conical bottom yellow wells
polypropylene
yellow plate
feature
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128516
technique(s)
PCR: suitable
size
384 wells
well maximum volume
40 μm
Legal Information
Eppendorf is a registered trademark of Eppendorf AG