Vybrat velikost
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 1 ea | K dispozici k odeslání DNESodAreál Kühne+Nagel spol. s r.o. | 11 800,00 Kč |
O této položce
11 800,00 Kč
K dispozici k odeslání DNESpodrobné informace
packaging
pkg of 1 pack
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable, electrophoresis: suitable
compatibility
for use with mPAGE® Mini Wet Transfer System
General description
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| compatibility for use with mPAGE® Mini Wet Transfer System | compatibility for use with mPAGE® Mini Wet Transfer System. | compatibility for use with mPAGE® Mini Gel Tank | compatibility for use with mPAGE® Mini Wet Transfer System |
| packaging pkg of 1 pack | packaging pkg of 1 pack | packaging pkg of 1 system, pkg of 1 ea ( for transferring up to 2 gels) | packaging pkg of 5 packs |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
| technique(s) SDS-PAGE: suitable, electrophoresis: suitable | technique(s) SDS-PAGE: suitable, electrophoresis: suitable | technique(s) immunoblotting: suitable, western blot: suitable | technique(s) immunoblotting: suitable, western blot: suitable |