Vybrat velikost
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 6 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 410,00 Kč |
O této položce
5 410,00 Kč
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
feature
needle type F (cemented needle)
packaging
pkg of 6 ea
manufacturer/tradename
SGE 002814
needle L
42 mm
volume
10 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
General description
1 of 1
Tato položka | |||
|---|---|---|---|
| packaging pkg of 6 ea | packaging pkg of 6 ea | packaging pkg of 6 ea | packaging pkg of 6 ea |
| material cone tip stainless steel needle (point style 1) | material cone tip stainless steel needle (point style 1) | material cone tip stainless steel needle (point style 1) | material cone tip stainless steel needle (point style 1) |
| manufacturer/tradename SGE 002814 | manufacturer/tradename SGE | manufacturer/tradename SGE 001814 | manufacturer/tradename SGE 001804 |
| volume 10 μL | volume 10 μL | volume 5 μL | volume 5 μL |
| compatibility for use with Agilent/HP 7673 and 7683 autosampler | compatibility for use with Agilent/HP 7673 and 7683 autosampler | compatibility for use with Agilent/HP 7673 and 7683 autosampler | compatibility for use with Agilent/HP 7673 and 7683 autosam pler |
| needle size 23 ga | needle size 26 ga | needle size 23 ga | needle size 26 ga |
Still not finding the right product?
Explore all of our products under SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| 21544 | 04061833036587 |