Hlavní dokumenty
21911
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 5 μL, SK-5F-HP-0.63, needle size 23 ga, needle L 42 mm, pkg of 6 ea
About This Item
UNSPSC Code:
41122001
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
feature
needle type F (cemented needle)
packaging
pkg of 6 ea
manufacturer/tradename
SGE 001814
needle L
42 mm
volume
5 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)