Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 pkg | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 710,00 Kč |
3 pkg | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 120,00 Kč |
O této položce
1 710,00 Kč
material
polypropylene bottle, polypropylene cap, round translucent bottle
joint
Joints threaded neck (70)
agency
suitable for EPA OTM-45
description
Autoclavable with closure completely unscrewed
feature
with cap
packaging
pkg of 12
manufacturer/tradename
Nalgene® 2118-0004
capacity
125 mL
application(s)
PFAS testing
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| material polypropylene bottle, round translucent bottle, polypropylene cap | material polypropylene bottle, polypropylene cap, round translucent bottle | material polypropylene bottle, polypropylene cap, round translucent bottle | material polypropylene bottle, polypropylene cap, round translucent bottle |
| feature with cap | feature with cap | feature with cap | feature with cap |
| manufacturer/tradename Nalgene® 2118-0004 | manufacturer/tradename Nalgene® 2118-0032 | manufacturer/tradename Nalgene® 2118-0008 | manufacturer/tradename Nalgene® 2118-0002 |
| capacity 125 mL | capacity 1000 mL | capacity 250 mL | capacity 60 mL |
| joint Joints threaded neck (70) | joint Joints threaded neck (120) | joint Joints threaded neck (70) | joint Joints threaded neck (53) |
| packaging pkg of 12 | packaging pkg of 6 | packaging pkg of 6 | packaging pkg of 12 |
Still not finding the right product?
Explore all of our products under Jars Nalgene® style 2118
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?