Změnit zobrazení
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 12 900,00 Kč |
O této položce
NACRES:
NB.01
UNSPSC Code:
41121500
12 900,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlifeature
adjustable, single-channel
manufacturer/tradename
Heathrow Scientific 120554
volume × volume × volume
2-20 μL × 20-200 μL × 100-1000 μL
General description
The Pipette Starter Kit is convenient for new labs or for customers who want a new set of pipettors covering the most commonly used volumes. The kit includes 3 single channel Pearl Pipettes, a Universal Carousel Pipette Stand and five Reagent Reservoirs–all at a value price!
Features and Benefits
Universal Carousel Pipette Stand (120499):
- Holds up to eight single-channel, four multi-channel, or a combo of both pipettes
- Weighted base allows for easy removal or replacement of pipettes, with one hand
- Pipettes are held in a vertical position to help prevent contamination
- Conveniently rotate stand head to select or store pipettes
- Quick grip handle to move pipette stand (loaded or unloaded)
- Assorted color options allow for color coding pipette location
- The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
- U.S. Patent Pending
- Internal metal parts for increased accuracy and calibration stability, and long life
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digital volume display
- Labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified
- Autoclavable
- Tapered reservoir bottoms form into a narrow trough to aid full sample recovery
- Disposable
- Sterile
Legal Information
Pearl is a trademark of Specac, Inc.
1 of 1
Tato položka | |||
|---|---|---|---|
| feature adjustable, single-channel | feature single-channel | feature Holds 1 x pipetors, for 1 Eppendorf® Xplorer® or Xplorer® plus, for Charger Carousel 2 | feature Holds 1 x pipetors, for 1 Eppendorf Xplorer® or Xplorer® plus |
| volume × volume × volume 2-20 μL × 20-200 μL × 100-1000 μL | volume × volume × volume - | volume × volume × volume - | volume × volume × volume - |
| manufacturer/tradename Heathrow Scientific 120554 | manufacturer/tradename Corning | manufacturer/tradename Eppendorf® 3116602007 (Global) | manufacturer/tradename Eppendorf® 3116000031 (Global) |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Související obsah
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| HS120554-1EA | 04065265819336 |