O této položce
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Eppendorf® 3116602007 (Global)
Feature:
Holds 1 x pipetors, for 1 Eppendorf® Xplorer® or Xplorer® plus, for Charger Carousel 2
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlifeature
Holds 1 x pipetors, for 1 Eppendorf® Xplorer® or Xplorer® plus, for Charger Carousel 2
manufacturer/tradename
Eppendorf® 3116602007 (Global)
General description
Keep all your Eppendorf pipettes within good reach and save precious bench space. The new Eppendorf Pipette Holder System provides highly flexible carousels, stands and wall mount devices for all current Eppendorf liquid handling instruments and most predecessors.
The Eppendorf pipette holder system provides safe and convenient storage. As all pipettes are positioned upright, their nose cone does not touch the lab bench reducing the risk of damage and cross contamination. The carousels provide easy access to the pipette of your choice and have a robust design with a small footprint. They can hold up to six liquid handling instruments. The stands carry one electronic pipette or Multipette. Carousels and stands are available with and without charging functionality. The wall mount devices store one manual or electronic pipette or Multipette.
Features and Benefits
New Eppendorf Pipette Holder System stores all current manual and electronic Eppendorf pipettes (including the Multipette) and most of their predecessors
Charger carousel with intelligent charging electronics and increased capacity for up to six instruments
Rotatable holder with completely new design for all manual pipettes including Eppendorf Research®, Eppendorf Research® plus, Reference®, Reference® 2 and Biomaster®
Carousels and stands with interchangeable pipette holders and charger shells, no tools required
Large rubber feet protect carousels and stands from liquids spilled on bench top
Pictograms on all charger shells and pipette holders for clear assignment in the lab
Charger carousel with intelligent charging electronics and increased capacity for up to six instruments
Rotatable holder with completely new design for all manual pipettes including Eppendorf Research®, Eppendorf Research® plus, Reference®, Reference® 2 and Biomaster®
Carousels and stands with interchangeable pipette holders and charger shells, no tools required
Large rubber feet protect carousels and stands from liquids spilled on bench top
Pictograms on all charger shells and pipette holders for clear assignment in the lab
Legal Information
Biomaster is a registered trademark of Eppendorf SE
Eppendorf is a registered trademark of Eppendorf SE
Eppendorf Research is a registered trademark of Eppendorf SE
Reference is a registered trademark of Eppendorf SE
Xplorer is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Eppendorf® Pipette Holder System
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?