Vybrat velikost
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 27 700,00 Kč |
O této položce
27 700,00 Kč
feature
for pipette tips 300 ul, multifunction rocker switch
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 4861000147
volume
15-300 μL
color
orange
General description
- Multi-functional rocker with up is up and down is down functionality
- Selection dial for quick and easy selection of functions
- Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
- Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
- Powerful rechargeable battery for up to 8 hours work without the need for recharging
- Separate power socket: Work can continue while charging
- Adjustment to external conditions (e.g. geographic height, liquids with high density)
- Lower part of pipette can be autoclaved to ensure decontamination
- Spring-loaded tip cone (not for 1,200?μL multi-channel)
- Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| feature for pipette tips 300 ul, multifunction rocker switch | feature for pipette tips 200 ul, multifunction rocker switch | feature for pipette tips 1250 ul, multifunction rocker switch | feature for pipette tips 300 ul, multifunction rocker switch |
| volume 15-300 μL | volume 5-100 μL | volume 50-1200 μL | volume 15-300 μL |
| manufacturer/tradename Eppendorf® 4861000147 | manufacturer/tradename Eppendorf® 4861000120 | manufacturer/tradename Eppendorf® 4861000163 | manufacturer/tradename Eppendorf® 4861000805 |
| color orange | color yellow | color green | color orange |
| packaging case of 1 ea | packaging case of 1 ea | packaging case of 1 ea | packaging case of 1 ea |
Still not finding the right product?
Explore all of our products under Eppendorf® Xplorer® 8-channel pipettor
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| EP4861000147-1EA | 04061841436317 |