Vybrat velikost
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 20 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 970,00 Kč 1 674,50 Kč |
O této položce
1 674,50 Kč
Ceníková cena1 970,00 KčUložit 15 %feature
Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths.
manufacturer/tradename
WHEATON®
Features and Benefits
- Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| manufacturer/tradename WHEATON® | manufacturer/tradename WHEATON® 900470 | manufacturer/tradename WHEATON® 900620 | manufacturer/tradename WHEATON® 900570 |
| feature Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths. | feature capacity 5 3 × 1 in. (76 × 25 mm) slide (vertically, ten slides back-to-back), Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths. | feature capacity 8 3 × 1 in. (76 × 25 mm) slide (sixteen slides back-to-back), Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths. | feature Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths. |
Still not finding the right product?
Explore all of our products under Wheaton Coplin staining jars
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| S5392-20EA | 04061826747223 |