Změnit zobrazení
| Velikost balení | Skladová položka | Dostupnost | Cena |
|---|---|---|---|
| 1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 860,00 Kč |
O této položce
UNSPSC Code:
41122001
NACRES:
NB.03
Material:
PTFE valve body
2 860,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
PTFE valve body
feature
positions 2
compatibility
for use with Hamilton PTFE Luer Lock (TLL) and other Luer lock syringes.
General description
This miniature, two-way valve from Hamilton provides flexibility, inertness, and small size for collecting gas samples and transporting or storing them in the original sampling syringe.
Fluid path is made entirely of PTFE to ensure inertness. Can be autoclaved for applications requiring sterility. 100 psig maximum pressure. Fits Hamilton PTFE Luer lock (TLL) and other Luer lock syringes.
Fluid path is made entirely of PTFE to ensure inertness. Can be autoclaved for applications requiring sterility. 100 psig maximum pressure. Fits Hamilton PTFE Luer lock (TLL) and other Luer lock syringes.
- Lever-actuated on/off valve for storing and transporting samples.
- KEL-F hub needles attach to the valve
- Bore ID 0.39"
Application
PTFE body two-way valve has been used in instrumental analysis of Cloud-point polyethylene glycol for imaging of immobilised single molecules using total-internal-reflection microscopy.
1 of 1
Tato položka | |||
|---|---|---|---|
| material PTFE valve body | material white PTFE valve | material PTFE Mininert valve | material PTFE Mininert valve |
| compatibility for use with Hamilton PTFE Luer Lock (TLL) and other Luer lock syringes. | compatibility for use with 20/400 mm thread | compatibility for use with VICI Series C and D Pressure-Lok syringes | compatibility for use with Hamilton TLL syringes |
| feature positions 2 | feature - | feature - | feature - |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Cloud-point PEG Glass Surfaces for Imaging of Immobilized Single Molecules by Total-internal-reflection Microscopy
Zhang, Zhengjian, et al.
Bio-protocol, 6, 2331-8325 (2016)
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| 20926 | 04061833406366 |