Zmień widok
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
| 1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 443,00 zł |
Informacje o tej pozycji
eCl@ss:
32070500
UNSPSC Code:
41122001
NACRES:
NB.03
443,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81027
needle L
, n/a
volume
100 μL
fitting
luer lock (without slots)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used in controlling the flow rate of the liquid samples and mobile phase during ion-pair solvent extraction performed in a microchannel fabricated in a quartz glass chip.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material PTFE Luer lock (with slots) | material PTFE Luer lock (with slots) | material PTFE Luer lock | material PTFE Luer lock |
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 81027 | manufacturer/tradename Hamilton 81127 | manufacturer/tradename Hamilton 80920 | manufacturer/tradename Hamilton 81020 |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
| volume 100 μL | volume 250 μL | volume 50 μL | volume 100 μL |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Pharmacology for the Prehospital Professional, 54-54 (2009)
Integration of a microextraction system on a glass chip: ion-pair solvent extraction of Fe (II) with 4, 7-diphenyl-1, 10-phenanthrolinedisulfonic acid and tri-n-octylmethylammonium chloride.
Tokeshi, Manabu, Tomoko Minagawa, and Takehiko Kitamori.
Analytical Chemistry, 72.7, 1711-1714 (2000)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| 26205 | 04061832873893 |