Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 345,00 zł |
Informacje o tej pozycji
eCl@ss:
32070500
UNSPSC Code:
41122001
NACRES:
NB.03
345,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80920
needle L
, n/a
volume
50 μL
fitting
luer lock (without slots)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.1 The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used during measuring phosphine concentration by gas chromatographic-flame photometric detector from the gas collected in fumigation chamber.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) |
| material PTFE Luer lock | material PTFE Luer lock | material PTFE Luer lock (with slots) | material PTFE Luer lock (with slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| volume 50 μL | volume 100 μL | volume 250 μL | volume 100 μL |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
| manufacturer/tradename Hamilton 80920 | manufacturer/tradename Hamilton 81020 | manufacturer/tradename Hamilton 81127 | manufacturer/tradename Hamilton 81027 |
Still not finding the right product?
Explore all of our products under Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?