Przejdź do
26206
Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock
1725TLL, volume 250 μL, PTFE Luer lock (with slots), needle size (not included)
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
358,00 zł
Informacje o tej pozycji
eCl@ss:
32070500
UNSPSC Code:
41122001
NACRES:
NB.03
358,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Zamów zamówienie zbiorcze
Pomoc techniczna
Pozwól nam pomóc
material
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81127
needle L
, n/a
volume
250 μL
fitting
luer lock (without slots)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been equipped in precision syringe pump of isothermal titration calorimeter which has been used in measuring the molar enthalpies of binary mixtures containing 2-decanone or dipentyl ether. It has also been used in the synthesis of single-molecule DNA by DNA polymerase with microflow channels.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
209972099920915
fitting
luer lock (without slots)
fitting
luer lock (without slots)
fitting
luer lock (without slots)
fitting
luer lock (without slots)
material
PTFE Luer lock (with slots)
material
PTFE Luer lock
material
PTFE Luer lock
material
PTFE Luer lock
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
needle L
, n/a
needle L
, n/a
needle L
, n/a
needle L
, n/a
needle size
(not included)
needle size
(not included)
needle size
(not included)
needle size
(not included)
manufacturer/tradename
Hamilton 81127
manufacturer/tradename
Hamilton 81320
manufacturer/tradename
Hamilton 81520
manufacturer/tradename
Hamilton 81120
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)