Wybierz wielkość
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
| 5 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 425,00 zł |
Informacje o tej pozycji
425,00 zł
material
blue , polypropylene
feature
to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific 120684
L × W × H
232 mm × 70 mm × 95 mm
General description
Features and Benefits
- Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection & culture tubes
- Only 4-way rack available with a six way connection, link different or the same well sizes, end to end or side by side, in any orientation
- Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit
- Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
- Molded alphanumeric identification for quick location and retrieval of tubes
- Robust, made from polypropylene that is chemical resistant and easy to clean
- Racks can be used in water baths and placed in freezers
- Assorted color pack available for color coding options
- Patent Pending
- Autoclavable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| manufacturer/tradename Heathrow Scientific 120684 | manufacturer/tradename Heathrow Scientific 120686 | manufacturer/tradename Heathrow Scientific 120687 | manufacturer/tradename Heathrow Scientific 120685 |
| material blue , polypropylene | material green , polypropylene | material clear , polypropylene | material polypropylene , purple |
| packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea |
| L × W × H 232 mm × 70 mm × 95 mm | L × W × H 232 mm × 70 mm × 95 mm | L × W × H 232 mm × 70 mm × 95 mm | L × W × H 232 mm × 70 mm × 95 mm |
| feature to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes | feature to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes | feature to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes | feature to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes |
Still not finding the right product?
Explore all of our products under Clinical 4-Way Tube Rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| HS120684-5EA | 04065265819701 |