Wybierz wielkość
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
| 100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 12 250,00 zł 10 412,50 zł |
Informacje o tej pozycji
10 412,50 zł
Cena katalogowa12 250,00 złZaoszczędź 15%material
black plate, black polystyrene , clear bottom, flat bottom
sterility
sterile
feature
lid, skirt, Low Flange, with generic barcode
packaging
case of 100 plates, pack of 20
manufacturer/tradename
Corning 3770BC
size
384 wells
well volume
112 μL
well working volume
25-50 μL
wells
384 wells
suitability
suitable for (cell cuture)
binding type
CellBIND® surface
Features and Benefits
- A flat, optically clear well bottom enables direct microscopic examination
- Low background fluorescence and opaque walls minimize crosstalk between wells and light scattering
- Wells are identified using individual alphanumeric codes
- Gamma radiation sterilized and nonpyrogenic
- Corning® CellBIND® treated surface for better cell attachment
- Suitable for the bottom and top reading instruments
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| size 384 wells | size 384 wells | size 384 wells | size 384 wells |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| material black plate, clear bottom, black polystyrene , flat bottom | material clear polystyrene , flat bottom | material black plate (clear bottom), black polystyrene , flat bottom | material black bottom, black polystyrene , flat bottom |
| packaging case of 100 plates, pack of 20 | packaging pack of 100 | packaging case of 100, pack of 20 | packaging pack of 10 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| feature lid, Low Flange, skirt, with generic barcode | feature lid | feature lid, skirt, plate format: low flange, with lid | feature lid, skirt, with generic barcode |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| CLS3770BC-100EA | 04061833884485 |
| CLS3770BC-20EA | 04061833884492 |