Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 655,00 zł |
Informacje o tej pozycji
655,00 zł
material
PTFE Luer lock
needle size
(not included)
agency
EPA (designed for analyzing drinking water samples), suitable for EPA 601
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81520
needle L
, n/a
volume
5 mL
fitting
luer lock (without slots)
compatibility
for use with purge and trap (US EPA)
General description
Application
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material PTFE Luer lock | material PTFE Luer lock | material PTFE Luer lock (with slots) | material Kel-F CTFE Luer lock |
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| agency EPA (designed for analyzing drinking water samples), suitable for EPA 601 | agency - | agency - | agency - |
| manufacturer/tradename Hamilton 81520 | manufacturer/tradename Hamilton 81320 | manufacturer/tradename Hamilton 81327 | manufacturer/tradename Hamilton 86020 |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
Still not finding the right product?
Explore all of our products under Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?