Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 319,00 zł |
Informacje o tej pozycji
319,00 zł
material
cone tip stainless steel needle (point style 1)
needle size
23s ga (cone tip)
feature
needle type N (cemented needle)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80387
needle L
43 mm (1.71 in.)
volume
10 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
General description
The plunger is not to be used with any other syringe.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material cone tip stainless steel needle (point style 1) | material cone tip stainless steel needle (point style 1) | material cone tip needle (point style 1) | material cone tip stainless steel needle (point style 1) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 6 ea |
| manufacturer/tradename Hamilton 80387 | manufacturer/tradename Hamilton 80388 | manufacturer/tradename Hamilton 87987 | manufacturer/tradename Hamilton 80391 |
| needle L 43 mm (1.71 in.) | needle L 43 mm (1.71 in.) | needle L 43 mm (1.71 in.) | needle L 43 mm (1.71 in.) |
| compatibility for use with Agilent/HP 7673 and 7683 autosampler | compatibility for use with Agilent/HP 7673 and 7683 autosampler | compatibility - | compatibility for use with Agilent/HP 7673 and 7683 autosampler |
| needle size 23s ga (cone tip) | needle size 26s ga (cone tip) | needle size 23s ga (cone tip) | needle size 23s-26s ga (cone tip) |
Still not finding the right product?
Explore all of our products under Hamilton® Microliter™ syringe, cemented needle
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?