Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 560,00 zł |
Informacje o tej pozycji
eCl@ss:
32070500
UNSPSC Code:
41122001
NACRES:
NB.03
560,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81420
needle L
, n/a
volume
2.5 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1002TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used during calorimetric analysis for phthalate protonation in different ionic media.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting - | fitting luer lock (without slots) |
| material PTFE Luer lock | material Kel-F CTFE Luer lock | material Kel-F CTFE Luer lock | material PTFE Luer lock |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| compatibility for use with Hamilton 1002TLL (PTFE Luer lock) | compatibility - | compatibility - | compatibility for use with purge and trap (US EPA) |
| volume 2.5 mL | volume 50 mL | volume 25 mL | volume 5 mL |
| needle L , n/a | needle L , n/a | needle L , n/a | needle L , n/a |
Still not finding the right product?
Explore all of our products under Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?