Przejdź do
Z709077
ART® non-filtered tip
HLT 300U SOFTFIT, size 1-300 μL, Pure packed, non-sterile
Synonim(y):
pipet tips, pipette tips, MBP 3771, dispensing tip, liquid transfer tip
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 pkg
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
620,00 zł
558,00 zł
5 pkg
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
2650,00 zł
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
1—300 μL
Sterility:
non-sterile
Feature:
DNase free, RNase free, Pure packed
Packaging:
pack of 960 ea
Manufacturer/tradename:
Molecular BioProducts 3771
558,00 zł
Cena katalogowa620,00 złZaoszczędź 10%
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
sterility
non-sterile
feature
DNase free, RNase free, Pure packed
packaging
pack of 960 ea
manufacturer/tradename
Molecular BioProducts 3771
maximum volume
300 μL
size
1-300 μL
volume
1—300 μL
General description
Easily access pipette tips with one hand using the user-friendly ART® non-filtered tips. It is designed to match your workflow and avoid concerns about tip spills on your bench or floor.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Features and Benefits
  • Ensures quality, performance, and reliability.
  • Certified sterile tips are free of DNase, RNase, DNA, and pyrogen contamination.
  • Racks designed for convenient and efficient access to pipette tips, saving valuable time during experiments.
  • Front latch mechanism securely closes and locks, providing safe and protected storage of tips.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z709050Z709042Z709107
manufacturer/tradename
Molecular BioProducts 3771
manufacturer/tradename
Molecular BioProducts 3571
manufacturer/tradename
Molecular BioProducts 3552
manufacturer/tradename
Molecular BioProducts 3582
feature
DNase free, Pure packed, RNase free
feature
DNase free, RNase free, Pure packed
feature
DNase free, RNase free, racked
feature
DNase free, RNase free, racked
volume
1—300 μL
volume
1—300 μL
volume
0.5—200 μL
volume
1000 μL
sterility
non-sterile
sterility
sterile
sterility
non-sterile
sterility
non-sterile
packaging
pack of 960 ea
packaging
pack of 960 ea
packaging
pack of 960 ea
packaging
pack of 800 ea
maximum volume
300 μL
maximum volume
300 μL
maximum volume
200 μL
maximum volume
1000 μL
Still not finding the right product?
Explore all of our products under ART® non-filtered tip
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)