Zmień widok
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
| 1 pkg | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2490,00 zł |
Informacje o tej pozycji
NACRES:
NB.13
UNSPSC Code:
41161502
Material:
polystyrene chamber
Sterility:
sterile
Packaging:
pkg of carton of 16
2490,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
polystyrene chamber
sterility
sterile
feature
CE marked
manufacturer/tradename
Nunc 154852
packaging
pkg of carton of 16
well surface area
4.0 cm2
working volume
1.0-2.0 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek® II - CC2 Chamber Slide System
- Chemically coated growth surface on glass slide that mimics polylysine
- Provides binding sites optimal for fastidious cells (e.g. neurons)
- Growth surface remains stable without refrigeration
- Light blue frosted writing area
- Slide seperator included in each pack
- CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material polystyrene chamber | material colorless , polystyrene chamber | material colorless , polystyrene chamber | material polystyrene chamber |
| manufacturer/tradename Nunc 154852 | manufacturer/tradename Nunc 154941 | manufacturer/tradename Nunc 154917 | manufacturer/tradename Nunc 154534 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| packaging pkg of carton of 16 | packaging pkg of carton of 16 | packaging pkg of carton of 16 | packaging pack of 16 × |
| well surface area 4.0 cm2 | well surface area - | well surface area - | well surface area - |
| working volume 1.0-2.0 mL | working volume 0.2-0.5 mL | working volume 0.5-1.0 mL | working volume 0.2-0.5 mL |
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® II - CC2™ Chamber Slide™ system
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| S6565-1PAK | 04061834377030 |