Wybierz wielkość
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
| 1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 788,00 zł |
Informacje o tej pozycji
788,00 zł
material
epoxy-coated handle, low-density polyethylene
feature
Holds 1 x 4 L bottle
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 6501-4000
General description
Clear plastic cap protects bottle closure and allows identification of color-coded closure on bottles. Remove the cap with a twist and dispense without removing the bottle from the carrier.
Holds and protects standard acid and chemical bottles and permits pouring without removing the bottles.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material epoxy-coated handle, low-density polyethylene | material low-density polyethylene | material epoxy-coated handle, low-density polyethylene | material polypropylene cap, polypropylene copolymer bottle (PPCO), round translucent bottle, white polypropylene screw closure |
| manufacturer/tradename Nalgene® 6501-4000 | manufacturer/tradename Nalgene® 6505-0010 | manufacturer/tradename Nalgene® 6501-2500 | manufacturer/tradename Nalgene® 2126-4000 |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| feature Holds 1 x 4 L bottle | feature Holds 6 x 0.5 L bottle | feature Holds 1 x 2.5 L bottle | feature with cap |
Still not finding the right product?
Explore all of our products under Bottle carrier, Nalgene®
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Powiązane treści
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| C2062-1EA | 04061826744017 |