MilliporeSigma
Bottles & Cans
We offer a variety of packaging options for your application. Whether you need your solvent packaging in glass bottle, PVC coated bottle, Sure/Seal™ bottle or aluminum can, we package and solvent dispensing options to meet your needs.
Bottle/Packaging Specifications
For Amber Glass, PVC Coated, and Sure/Seal™ Bottles
CapacityBottle Dimension
(H x D) cm
ClosureCase PackCase Pack Dimension
(L x W x H) cm
100 mL11.91 x 5.5133-43012 x 100 mL34.93 x 24.13 x 24.77
500 mL18.75 x 8.8133-4306 x 500 mL34.93 x 24.13 x 24.77
1 L22.86 x 10.3433-4306 x 1 L39.37 x 26.83 x 27.94
2 L33.98 x 13.3338-4394 x 2 L33.78 x 37.78 x 40.00
4 L34.29 x 16.2038-4394 x 4 L33.78 x 37.78 x 40.00
Amber Glass Bottles
Our solvents are available in a variety of amber glass bottles. The inert composition of glass makes them ideal for storing and dispensing of high purity solvents.
PVC Coated Bottles
To help prevent chemical spills, we provide solvents in shatter-resistant, poly vinyl chloride coated bottles. The plastic coating aids in absorbing some of the shock in the event the bottle is dropped and assists in containing the spill. Selected solvents are available in 100mL, 1L and 4L PVC coated bottles (refer to above Bottle/Packaging Specifications).
Pumping
Pumping
Hirschmann® Ceramus® bottle-top dispenser (Z341479)
  • Chemically inert − unique ceramic dispenser piston in a Duran borosilicate glass barrel
  • Micro-step volume adjustment − ensures reproducibility
  • Rotates on the bottle − for convenience
  • Recirculation system − for fast priming without loss of reagent
  • Lifetime calibration − supplied with individual serial number and Test Certificate
HPLC Delivery Caps
HPLC Delivery Caps
For direct solvent delivery to HPLC instruments or simple pressure transfers. Each Delivery Cap includes 10ft. of PFA tubing. Caps require a GL-45 bottle adapter for use with 2L and 4L bottles.
Two-hole Delivery Cap (Z543845)
Three-hole Delivery Cap (Z543586)
Glass Bottles Storage and Dispensing Options
Glass Bottles Storage and Dispensing Options
4L Bottle Jacket (Z256366)
Provides virtually unbreakable, secondary containment for transport and storage of chemicals. Meets OSHA Laboratory Standards (29CFR1910-1450).
  • Rugged PE construction
  • Solvent labels and levels clearly visible through jacket
  • Convenient, handle-locking mechanism keeps top and bottom of jacket locked together
Sure/Seal™ Bottles
Sure/Seal™ Bottles
The Sure/Seal™ bottle incorporates the inert composition of our amber glass bottles with a time-tested crimp-top system. This design ensures the exceptional state of dryness achieved at time of purification is maintained throughout long-term storage and use.
Before filling, low levels traces of moisture are removed from the bottle through oven drying. The bottles are then filled under an inert atmosphere of dry nitrogen to prevent moisture build-up and sealed with our unique Sure/Seal™ liner and crown cap. Anhydrous and Biotech grades are available in 100mL, 1L and 2L Sure/Seal™ bottles (refer to above Bottle/Packaging Specifications).
Dispensing Methods for Sure/Seal™ Bottles
Dispensing Methods for Sure/Seal™ Bottles
Standard syringe and cannula techniques are used to transfer the contents.
See Technical Bulletin AL-134.
Sure/Seal™ Transfer Accessories
CHEM-FLEX Transfer Line
Two 12-gauge SS needles (6 and 18 in.) are connected to the CHEM-FLEX 106 tubing with nylon clamps. Solvents contact only PTFE and SS during transfers.
Sure/Stor™ flasks
Sure/Stor™ Flasks
Sure/Stor™ Flasks are designed for safe, reliable storage and dispensing of air-sensitive and odoriferous chemicals, solvents, pyrophorics, alkyl lithiums, Grignards and corrosives.
Features:
  • High-vacuum PTFE valve-to-glass seal eliminates air contamination in storage and septum leakage after initial needle penetration
  • Heavy-wall borosilicate glass
  • Removable side-arm hose connector for easy attachment of vacuum and inert gas lines. For long-term storage, replace hose connector with a rubber septum for secondary protection.
Aluminum Bottles
Aluminum Bottles
These one-piece seamless aluminum bottles are corrosion-resistant and U.N. approved.
  • Pure 1050A aluminum (min. 99.5%)
  • Polypropylene closure with PTFE liner
  • Anti-back off, tamper evident locking closure
  • Easily recyclable
CapacityBottle Dimension (H x D) cmClosureCase PackCase Pack Dimension (L x W x H) cm
1 L24.3 x 8.832 mm6 x 1 L39.37 x 26.83 x 27.94
4 L23.9 x 17.545 mm4 x 4 L36.20 x 36.20 x 39.37
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch