Przejdź do
26284
Hamilton® SampleLock syringe
1001SL, volume 1 mL, needle size 22 ga (bevel tip), needle L 51 mm (2 in.)
Wybierz wielkość
Zmień widok
Gabaryty przesyłkiSKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1090,00 zł
Informacje o tej pozycji
UNSPSC Code:
41122001
NACRES:
NB.03
1090,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Zamów zamówienie zbiorcze
Pomoc techniczna
Pozwól nam pomóc
material
bevel tip stainless steel needle (point style 2)
needle size
22 ga (bevel tip)
feature
syringe type Sample Lock
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81356
needle L
51 mm (2 in.)
volume
1 mL
General description
  • Twist valve locks the sample in the syringe
  • Positive rear plunger stop eliminates sample loss and plunger blowout at high pressures (not applicable to 1705 and 1710 models)
  • Connects to either male or female luer fittings by using an adapter
  • Male and female luer and male luer lock adapters thread onto the SampleLock valve making the syringe compatible with many connectors and fittings
  • All adapters have a Kel-F (CTFE) fluid path, with a port diameter of 0.039 in.
  • Internal standards can be added through the valve port into the sample contained in the syringe
Application
Hamilton® SampleLock syringe has been used in sampling gas and liquid phase for GC analysis, in isothermal vapor-liquid equilibrium data measured for two binary systems.
Other Notes
We recommend replacement needle Cat. No. 19136-U (22s ga needle) for the 1705SL, 1710SL and 1725SL syringes; Cat. No. 19137-U (22 ga needle) is recommended for the larger syringes. Both replacement needles will fit 250 μL to 10 mL syringes and will fit any SampleLock syringe.
Plunger Assemblies are available for the 1705SL (Cat. No. 26152-U) and 1710SL (Cat. No. 26154) syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
262812628326285
material
bevel tip stainless steel needle (point style 2)
material
bevel tip stainless steel needle (point style 2)
material
bevel tip stainless steel needle (point style 2)
material
bevel tip stainless steel needle (point style 2)
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
needle L
51 mm (2 in.)
needle L
51 mm (2 in.)
needle L
51 mm (2 in.)
needle L
51 mm (2 in.)
volume
1 mL
volume
100 μL
volume
0.5 mL
volume
2.5 mL
manufacturer/tradename
Hamilton 81356
manufacturer/tradename
Hamilton 81056
manufacturer/tradename
Hamilton 81256
manufacturer/tradename
Hamilton 81456
feature
syringe type Sample Lock
feature
syringe type Sample Lock
feature
syringe type Sample Lock
feature
syringe type Sample Lock
Still not finding the right product?
Explore all of our products under Hamilton® SampleLock syringe
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Vapor- liquid equilibrium for the binary systems of 3-methylpentane+ 2-methyl-2-propanol at 331 K and+ 2-butanol at 331 K.
Uusi-Kyyny, Petri, et al.
Journal of Chemical and Engineering Data, 46.3, 754-758 (2001)
Numer pozycji handlu globalnego
SKUNUMER GTIN
1544949-200MG04061833832417
P215501004061833814055
PHR3090-400MG04065266900149
413283-100G04061833597835
2628404061832873770