Wybierz wielkość
| Gabaryty przesyłki | SKU | Dostępność | Cena netto |
|---|---|---|---|
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 5470,00 zł |
Informacje o tej pozycji
5470,00 zł
Quality Level
description
Cell line type: Embryonic Stem Cells, Mouse Embryonic Stem Cells
technique(s)
cell culture | stem cell: suitable
General description
RASA: C3H/HeN
PŁEĆ: Mężczyzna
KARYOTYP: 40XY
Packaging
Physical form
Preparation Note
Other Notes
Disclaimer
Certyfikaty analizy (CoA)
Poszukaj Certyfikaty analizy (CoA), wpisując numer partii/serii produktów. Numery serii i partii można znaleźć na etykiecie produktu po słowach „seria” lub „partia”.
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.
Powiązane treści
"As transgenic mouse core laboratories know well, mouse models are a powerful means of determining the function of mammalian genes in the context of whole organisms—especially since over 95% of the mouse genome is similar to the human genome. However, you may not know that: 1. Laboratory mouse strains are descended from ancestors in modern-day Pakistan. 2. The first inbred strain of lab mouse was created in 1909. It was named DBA for its coat color: dilute brown non-agouti. 3. There is a mutant mouse actually named“Bad Hair Day” (Bhrd). Someone get this little guy a stylist! 4. Male mice, when treated with anticancer drugs, have been shown to pass on DNA instability to their offspring. 5. The founder of Jackson Laboratories, C. C. Little, once had lunch with Walt Disney to request that an animated film be created linking Mickey Mouse and the laboratory mouse. The request was denied."
The cell culture protocols described in this instructional manual include the in vitro culture of murine ES cells using EmbryoMax products along with ESGRO® mLIF medium supplement, as well as feeder-free and serum-free ES cell culture using the ESGRO Complete™ line of products. Also included in this instructional manual are: methods for serum-free neuronal differentiation of mouse ES cells, iPS cell generation using STEMCCA™ lentivirus kits, cre-recombinase mediated excision of STEMCCA™ genes from iPS cells, derivation and rescue of new and existing ES cell lines using RESGRO™ Culture Medium and ESGRO Complete™ system.