material
colorless
sterility
non-sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
manufacturer/tradename
Sartorius 791002MAX
packaging
pack of 960 ea (Refil), pkg of (refill pack)
maximum volume
1 mL
volume range
10-1,000 μL
volume
10—1000 μL
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| volume 10—1000 μL | volume 0.5—200 μL | volume 0.1—10 μL | volume 5—350 μL |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| feature DNase free, barrier: no, RNase free, endotoxin free | feature DNase free, RNase free, barrier: no, endotoxin free | feature DNase free, RNase free, barrier: no, endotoxin free | feature DNase free, RNase free, barrier: no, endotoxin free |
| manufacturer/tradename Sartorius 791002MAX | manufacturer/tradename Sartorius 790202MAX | manufacturer/tradename Sartorius 790012MAX | manufacturer/tradename Sartorius 790350 |
| material colorless | material colorless | material colorless | material colorless |
| packaging pack of 960 ea (Refil), pkg of (refill pack) | packaging pack of 960 ea (Refil), pkg of (refill Tower) | packaging pack of 960 ea (Refil), pkg of (refill Tower) | packaging pack of 96 ea (Racked) |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| Z741673-9600EA | 04061832831879 |