feature
to hold 12 x 1122, 1127-A, 1621 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1628
parameter
2,683 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Universal 320/320R and Rotafix 32A
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| compatibility for use with centrifuge models Universal 320/320R and Rotafix 32A | compatibility for use with centrifuge models ROTANTA 460/460R | compatibility for use with centrifuge models Universal 320/320R and Rotafix 32A | compatibility for use with centrifuge models Universal 320/320R and Rotafix 32A |
| manufacturer/tradename Hettich 1628 | manufacturer/tradename Hettich 4446 | manufacturer/tradename Hettich 1619 | manufacturer/tradename Hettich 1617 |
| parameter 2,683 max. RCF | parameter 3,631 max. RCF | parameter 2,701 max. RCF | parameter 2,469 max. RCF |
| feature to hold 12 x 1122, 1127-A, 1621 buckets | feature capacity 1.5 ‑ 100 mL, to hold 6 x 5092 buckets | feature to hold 6 x 50 mL tubes with screw caps | feature to hold 8 x 50 mL tubes with screw caps |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?