Przejdź do
55222
6-Carboxytetramethylrhodamine
BioReagent, suitable for fluorescence, ≥90% (HPLC)
Synonim(y):
6-TAMRA
Wybierz wielkość
Zmień widok
Gabaryty przesyłkiSKUDostępnośćCena netto
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C25H22N2O5
Numer CAS:
Masa cząsteczkowa:
430.45
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Aby zapytać o ten produkt 55222, skontaktuj się z lokalnym biurem lub sprzedawcą firmy Merck. Skontaktuj się z zespołem ds. pomocy technicznej
Pomoc techniczna
Pozwól nam pomóc
description
single isomer
product line
BioReagent
assay
≥90% (HPLC)
form
solid
solubility
DMSO: soluble, methanol: soluble
fluorescence
λex 543 nm; λem 575 nm in 0.1 M Tris pH 8.0
suitability
suitable for fluorescence
SMILES string
CN(C)c1ccc2c(OC3=C\C(C=CC3=C2c4cc(ccc4C([O-])=O)C(O)=O)=[N+](\C)C)c1
InChI
1S/C25H22N2O5/c1-26(2)15-6-9-18-21(12-15)32-22-13-16(27(3)4)7-10-19(22)23(18)20-11-14(24(28)29)5-8-17(20)25(30)31/h5-13H,1-4H3,(H-,28,29,30,31)
InChI key
COCMHKNAGZHBDZ-UHFFFAOYSA-N
Application
6-Carboxytetramethylrhodamine (6-TAMRA) is used to create fluorescent DNA probes and labeled proteins/peptides. The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.
The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Klasa składowania
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Kevin J Landmark et al.
ACS nano, 2(4), 773-783 (2009-02-12)
Organic-coated superparamagnetic iron oxide nanoparticles (OC-SPIONs) were synthesized and characterized by transmission electron microscopy and X-ray photoelectron spectroscopy. OC-SPIONs were transferred from organic media into water using poly(amidoamine) dendrimers modified with 6-TAMRA fluorescent dye and folic acid molecules. The saturation
Patrick A Marcotte et al.
Analytical biochemistry, 332(1), 90-99 (2004-08-11)
A novel fluorescent substrate was devised for the sirtuin (SIRT) class of human protein deacetylases comprised of a peptide sequence containing a single acetyl-lysine residue, with a fluorescent group (tetramethylrhodamine-6-carboxylic acid, 6-TAMRA) near the carboxyl terminus and a nonfluorescent quenching
Redouane Bouchaala et al.
Colloids and surfaces. B, Biointerfaces, 156, 414-421 (2017-05-30)
Light is an attractive trigger for release of active molecules from nanocarriers in biological systems. Here, we describe a phenomenon of light-induced release of a fluorescent dye from lipid nano-droplets under visible light conditions. Using auto-emulsification process we prepared nanoemulsion
Numer pozycji handlu globalnego
SKUNUMER GTIN
M103-10MG04061832091785
55222-50MG-F04061832532578
55222-10MG-F04061832532561