Przejdź do zawartości
Merck

Przejdź do

MABE350

Anti-phospho-RNAPII (Tyr1) Antibody, clone 3D12

clone 3D12, from rat

Synonim(y):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Gabaryty przesyłkiSKUDostępnośćCena netto
100 μg

Dostępne do wysyłki DZISIAJzKuehne + Nagel Sp. z o.o.

2060,00 zł

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
3D12, monoclonal
Species reactivity:
human
Application:
ChIP, IP, WB
Citations:
1

2060,00 zł


Dostępne do wysyłki DZISIAJSzczegóły


Zamów zamówienie zbiorcze
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D12, monoclonal

species reactivity

human

technique(s)

ChIP: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1)

General description

RNA polymerase II (RNAPII or Pol II) is a multi-subunit enzyme responsible for the transcription of transcription of DNA into RNA from protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. RNAPII can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. The DNA binding domain of the polymerase is a groove where TFIIB orients the DNA for unwinding and transcription.
~220 kDa observed

Immunogen

Ovalbumin-conjugated linear peptide corresponding to human RNAPII phosphorylated at Tyr1.

Application

Detect the phospho-RNAPII (Tyr1) protein using this Anti-phospho-RNAPII (Tyr1), clone 3D12 validated for use in WB, IP & ChIP.
Immunoprecipitation Analysis: A representative lot from an independent laboratory detected phospho-RNAPII (Tyr1) in proliferating yeast whole cell extract (Mayer, A., et al. (2012). Science. 336(6089):1723-1725.).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot from an independent laboratory immunoprecipitated phospho-RNAPII (Tyr1) in proliferating yeast whole cell extract (Mayer, A., et al. (2012). Science. 336(6089):1723-1725.).

Physical form

Format: Purified

Analysis Note

Control
Untreated and lambda phosphatase treated HeLa cell lysates
Evaluated by Western Blot Analysis in untreated and lambda phosphatase treated HeLa cell lysates.

Western Blot Analysis: 2 µg/mL of this antibody detected phospho-RNAPII (Tyr1) in 10 µg of untreated HeLa cell lysate. (Lambda phosphatase treatment was performed during the western blot procedure.)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
ZRB1409MABE348MABE954
species reactivity

human

species reactivity

mouse, human, rat

species reactivity

mouse, yeast, human

species reactivity

canine, mouse, rat, human, monkey

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

clone

3D12, monoclonal

clone

recombinant monoclonal

clone

1G7, monoclonal

clone

1H4B6, monoclonal

biological source

rat

biological source

rabbit

biological source

rat

biological source

rat

UniProt accession no.

P24928

UniProt accession no.

P24928

UniProt accession no.

P24928

UniProt accession no.

P24928

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certyfikaty analizy (CoA)

Poszukaj Certyfikaty analizy (CoA), wpisując numer partii/serii produktów. Numery serii i partii można znaleźć na etykiecie produktu po słowach „seria” lub „partia”.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów



null



Numer pozycji handlu globalnego

SKUNUMER GTIN
MABE35004053252833007

Questions

Reviews

No rating value

Active Filters