Vai a
C4176
Casting clamps
For single-gel casting assembly
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Informazioni su questo articolo
NACRES:
NB.51
UNSPSC Code:
12352200
Material:
Nylon
material
Nylon
General description
Specifically designed for gel casting. Wide jaws provide uniform pressure on glass plate-gasket assembly. Single- and dual-gel casting clamps accommodate different thicknesses and are not interchangeable.
Set of three (3). One set (three clamps) is sufficient for 10 cm wide mini-gels; two sets (six clamps) are necessary for standard 16.5 cm wide gels.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Tatsuya Osaki et al.
Scientific reports, 7(1), 1897-1897 (2017-05-17)
Consistent cell preparation is a fundamental preliminary step for understanding complex cellular mechanisms in various cell-based research fields, including basic cell biology, cancer research, and tissue engineering. However, certain elusive factors, such as cellular de-differentiation and contamination with mycoplasma or
Numero articolo commerciale globale
SKUGTIN
C4176-1SET04061833491805