Fortfahren mit
S4767
Wheaton staining dishes
20 slides, covers only
Synonym(e):
900202
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
3 ea
Warenkorb auf Verfügbarkeit prüfen
CHF 116.00
CHF 98.60
Über diesen Artikel
NACRES:
MA.01
UNSPSC Code:
41100000
CHF 98.60
ListenpreisCHF 116.00Sparen Sie 15%
Aktion nur im Web
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
rectangular , soda-lime glass
packaging
pack of 3 ea
manufacturer/tradename
WHEATON® 900202
dish L × W
95 mm × 76 mm
dish capacity
3 x 1 in. (75 x 25 mm) , 10 single slides, 3 x 1-1/2 in. (75 x 38 mm) , 19 slides arranged alternately straight across and diagonally, 3 x 2 in. (75 x 51 mm) , 20 slides back-to-back of standard size
General description
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
Features and Benefits
  • Manufactured from soda-lime glass
  • This 20-slide unit is the standard for manual staining procedures
  • The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3” x 1” (75 x 25mm), 3” x 1-1/2” (75 x 38mm) and 3” x 2” (75 x 51mm) sizes
  • Approximate inside bottom dimensions (mm): 91 L x 71 W x 60 D
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z710954S5142S5017
manufacturer/tradename
WHEATON® 900202
manufacturer/tradename
WHEATON® 900303
manufacturer/tradename
WHEATON® 900205
manufacturer/tradename
WHEATON® 900204
material
rectangular , soda-lime glass
material
soda-lime glass
material
rectangular , stainless steel
material
rectangular , soda-lime glass
packaging
pack of 3 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
-
dish L × W
95 mm × 76 mm
dish L × W
-
dish L × W
95 mm × 76 mm
dish L × W
-
dish capacity
3 x 1 in. (75 x 25 mm) , 10 single slides, 3 x 2 in. (75 x 51 mm) , 20 slides back-to-back of standard size
dish capacity
~700 mL
dish capacity
3 x 1 in. (75 x 25 mm) , 10 single slides, 3 x 1-1/2 in. (75 x 38 mm) , 19 slides arranged alternately straight across and diagonally, 3 x 2 in. (75 x 51 mm)
dish capacity
-
Still not finding the right product?
Explore all of our products under Wheaton staining dishes
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)