Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | CHF 36.70 |
Über diesen Artikel
NACRES:
MA.01
UNSPSC Code:
41116155
CHF 36.70
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
silicone
description
double-indexed (numbers and letters) allowing for two embedments in the same mold
cavity size × depth
7-14 mm -3 mm
size
76-76 mm , mold
tip size
-3 mm
color
maroon
General description
With cavities for the preparation of microscopy samples. Two variations are available. Maroon molds contain additives for chemical and thermal resistance; they are durable and economical. Clear molds are used where optical clarity is necessary for precise sample alignment; recommended for use with low-temperature UV-cured resins.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| color maroon | color clear | color clear | color - |
| tip size -3 mm | tip size -3 mm | tip size -4 mm | tip size - |
| cavity size × depth 7-14 mm -3 mm | cavity size × depth 7-14 mm -3 mm | cavity size × depth 7-14 mm -4 mm | cavity size × depth - |
| size 76-76 mm , mold | size 76-76 mm , mold | size 55-76 mm , mold | size - |
| description double-indexed (numbers and letters) allowing for two embedments in the same mold | description double-indexed (numbers and letters) allowing for two embedments in the same mold | description - | description - |
| material silicone | material silicone | material silicone | material - |
Still not finding the right product?
Explore all of our products under Embedding molds
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?