Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 pkg | Warenkorb auf Verfügbarkeit prüfen | CHF 357.00 |
Über diesen Artikel
CHF 357.00
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
packaging
pack of 16 ea, case of 96 ea
manufacturer/tradename
Nunc 154453
well area
8.6 cm2
wells
1
working volume
2.0-4.5 mL
suitability
suitable for (cell culture; attachment and growth of cells)
General description
Features and Benefits
- Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
- Tabbed polystyrene lid allows gas exchange with minimal evaporation.
- Removal tool separates media chamber from glass slide.
- Biocompatible adhesive hydrophobic well border is printed on slide.
- Slide features a white frosted writing area.
- Includes: Chamber removal tool.
- Sterile
- Packed in convenient stackable incubation trays.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename Nunc 154453 | manufacturer/tradename Nunc 154461 | manufacturer/tradename Nunc 154526 | manufacturer/tradename Nunc 154534 |
| material colorless , polystyrene chamber | material colorless , polystyrene chamber | material colorless , polystyrene chamber | material polystyrene chamber |
| wells 1 | wells 2 | wells 4 | wells 8 |
| packaging pack of 16 ea, case of 96 ea | packaging pack of 16 ea, case of 96 ea | packaging pack of 16 ea, case of 96 ea | packaging pack of 16 × |
| feature CE marked | feature CE marked | feature CE marked | feature CE marked |
| suitability suitable for (cell culture; attachment and growth of cells) | suitability suitable for (cell culture; attachment and growth of cells) | suitability suitable for (cell culture; attachment and growth of cells) | suitability suitable for (cell culture; attachment and growth of cells) |
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® II Chamber Slide™ System Kammerobjektträger
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C6307-1CS | 04061834373940 |
| C6307-1PAK | 04061834373957 |