Fortfahren mit
B7910
Nalgene® bottles, style 2126
capacity 4000 mL
Synonym(e):
nalgene bottle, nalgene bottles, Nalgene 2126-4000, laboratory container, receptical, vessel
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
CHF 83.20
Über diesen Artikel
NACRES:
NB.17
UNSPSC Code:
41121800
CHF 83.20
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polypropylene cap, polypropylene copolymer bottle (PPCO), round translucent bottle, white polypropylene screw closure
joint
Joints threaded neck (83B)
description
Autoclavable
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 2126-4000
bottle diam. × H
155 mm × 338 mm
capacity
4000 mL
General description
Round, heavy-duty, thick walled, wide-mouth bottles. Screw closure contains a thermoplastic elastomer (TPE) gasket.
Features and Benefits
  • Nalgene® wide-mouth bottles are ideally used as a waste aspirator bottle or as an autoclavable vessel for scale-up activities in cell culture applications.
  • Made of heavy-duty chemically resistant polypropylene copolymer
  • Thick wall, can withstand repeated room-temperature vacuum applications.
  • A leak-proof closure prevents sample loss.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
B8035B8160Z380407
feature
with cap
feature
with cap
feature
with cap
feature
with cap
manufacturer/tradename
Nalgene® 2126-4000
manufacturer/tradename
Nalgene® 2126-1000
manufacturer/tradename
Nalgene® 2126-2000
manufacturer/tradename
Nalgene® 2122-0010
material
polypropylene cap, round translucent bottle, polypropylene copolymer bottle (PPCO), white polypropylene screw closure
material
polypropylene cap, polypropylene copolymer bottle (PPCO), round natural bottle, white polypropylene screw closure
material
polypropylene cap, polypropylene copolymer bottle (PPCO), round bottle, white polypropylene screw closure
material
polypropylene cap, polypropylene copolymer , polypropylene copolymer bottle (PPCO), square , translucent
capacity
4000 mL
capacity
1000 mL
capacity
2000 mL
capacity
4,000 mL
packaging
pack of 1 ea
packaging
pack of 6 ea
packaging
pack of 2 ea
packaging
pack of × 6 ea
bottle diam. × H
155 mm × 338 mm
bottle diam. × H
93 mm × 226 mm
bottle diam. × H
119 mm × 216 mm
bottle diam. × H
142 mm × 282 mm
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2126
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)