Fortfahren mit
B7907
Nalgene® Flaschen, Stil 2002
high-density polyethylene, capacity 1,000 mL, with cap, translucent
Synonym(e):
HDPE bottles, nalgene bottle, nalgene bottles, Nalgene 2002-0032, laboratory container, vessel
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
6 ea
Warenkorb auf Verfügbarkeit prüfen
CHF 37.20
Über diesen Artikel
NACRES:
NB.17
UNSPSC Code:
41121800
CHF 37.20
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
joint
Joints threaded neck (38)
description
Not autoclavable
feature
with cap
packaging
pack of 6 ea
manufacturer/tradename
Nalgene® 2002-0032
capacity
1,000 mL
General description
Boston-Stil (runde) Eng-Öffnungs Flasche
Features and Benefits
  • Nalgene® narrow-mouth lab-quality bottles are ideal for storing and transporting liquids or dry chemicals in the lab.
  • Highly resistant to chemicals, these bottles are excellent for long-term use.
  • Includes linerless polypropylene leak-proof closures for preventing sample loss.
  • Uniform wall thickness protects against splitting or puncturing.
  • Bottles can withstand temperatures in a freezer up to -100°C.
  • Manufactured from non-toxic materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
B7532B3533B7782
material
high-density polyethylene , polypropylene screw closure, translucent , polypropylene cap, round bottle
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
feature
with cap
feature
with cap
feature
with cap
feature
with cap
manufacturer/tradename
Nalgene® 2002-0032
manufacturer/tradename
Nalgene® 2002-0002
manufacturer/tradename
Nalgene® 2002-0016
manufacturer/tradename
Nalgene® 2002-0008
capacity
1,000 mL
capacity
60 mL
capacity
500 mL
capacity
250 mL
joint
Joints threaded neck (38)
joint
Joints threaded neck (20)
joint
Joints threaded neck (28)
joint
Joints threaded neck (24)
packaging
pack of 6 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
Still not finding the right product?
Explore all of our products under Nalgene® Flaschen, Stil 2002
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)