Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80401
needle L
, n/a
volume
25 μL
compatibility
for use with Hamilton 702LT (Luer tip)
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material glass body | material glass body | material glass body | material glass body |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| compatibility for use with Hamilton 702LT (Luer tip) | compatibility for use with Hamilton 725LT (Luer tip) | compatibility for use with Hamilton 750LT (Luer tip) | compatibility for use with Hamilton 705LT (Luer tip) |
| manufacturer/tradename Hamilton 80401 | manufacturer/tradename Hamilton 80701 | manufacturer/tradename Hamilton 80801 | manufacturer/tradename Hamilton 80501 |
| feature without needle Luer tip | feature without needle Luer tip | feature without needle Luer tip | feature without needle Luer tip |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Luer-Spitze
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 24531 | 04061832873510 |