Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.03
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80701
needle L
, n/a
volume
250 μL
compatibility
for use with Hamilton 725LT (Luer tip)
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material glass body | material glass body | material glass body | material PTFE tip, glass body |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| needle L , n/a | needle L , n/a | needle L , n/a | needle L , n/a |
| manufacturer/tradename Hamilton 80701 | manufacturer/tradename Hamilton 80501 | manufacturer/tradename Hamilton 80801 | manufacturer/tradename Hamilton 80601 |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
| compatibility for use with Hamilton 725LT (Luer tip) | compatibility for use with Hamilton 705LT (Luer tip) | compatibility for use with Hamilton 750LT (Luer tip) | compatibility for use with Hamilton 710LT (Luer tip) |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Luer-Spitze
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20703 | 04061832873015 |