Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
blunt tip Kel-F needle (point style 3)
needle size
22 ga (blunt tip)
feature
KEL-F hub
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90134
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material blunt tip Kel-F needle (point style 3) | material blunt tip stainless steel needle (point style 3) | material blunt tip stainless steel needle (point style 3) | material blunt tip stainless steel needle (point style 3) |
| needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 70 mm |
| packaging pkg of 6 ea | packaging pkg of 6 ea | packaging pkg of 6 ea | packaging pkg of 1 ea |
| feature KEL-F hub | feature needle blunt tip, Kel-F Hub (KF) | feature needle type Metal hub for TLL syringes, LT or TLL connectors | feature needle type LTN (fixed) |
| compatibility for use with Hamilton LT and TLL | compatibility for use with Hamilton LT and TLL | compatibility for use with Hamilton LT and TLL | compatibility for use with PerkinElmer GC Autosystem |
| manufacturer/tradename Hamilton 90134 | manufacturer/tradename Hamilton 90534 | manufacturer/tradename Hamilton 91022 | manufacturer/tradename Hamilton 88035 |
Still not finding the right product?
Explore all of our products under Hamilton® Kanülen für LT- und TLL-Spritzen
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Verwandter Inhalt
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 900406-500MG | 04061833244517 |
| 96408-10MG | 04061838445087 |
| 96408-50MG | 04061838445100 |
| 96408-250MG | 04061838445094 |
| 21744 | 04061833406458 |