Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
stainless steel needle (point style 2)
needle size
22s ga (bevel tip)
feature
removable needle RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 84859
needle L
51 mm (2 in.)
volume
100 μL
compatibility
for use with Hamilton 810RN (removable needle)
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material stainless steel needle (point style 2) | material stainless steel needle (point style 2) | material stainless steel needle (point style 2) | material stainless steel needle (point style 2) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| needle size 22s ga (bevel tip) | needle size 26s ga (bevel tip) | needle size 22s ga (bevel tip) | needle size 22s ga (bevel tip) |
| volume 100 μL | volume 10 μL | volume 25 μL | volume 50 μL |
| feature removable needle RN | feature removable needle RN | feature removable needle RN | feature removable needle RN |
| manufacturer/tradename Hamilton 84859 | manufacturer/tradename Hamilton 84853 | manufacturer/tradename Hamilton 84855 | manufacturer/tradename Hamilton 84857 |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Serie 800, abnehmbare Nadel
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 21498 | 04061832872858 |