Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
stainless steel needle (point style 2)
needle size
22s ga (bevel tip)
feature
removable needle RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 84857
needle L
51 mm (2 in.)
volume
50 μL
compatibility
for use with Hamilton 805RN (removable needle)
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material stainless steel needle (point style 2) | material stainless steel needle (point style 2) | material stainless steel needle (point style 2) | material stainless steel needle (point style 2) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| compatibility for use with Hamilton 805RN (removable needle) | compatibility - | compatibility for use with Hamilton 802RN (removable needle) | compatibility for use with Hamilton 810RN (removable needle) |
| needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 51 mm (2 in.) | needle L 51 mm (2 in.) |
| needle size 22s ga (bevel tip) | needle size 26s ga (bevel tip) | needle size 22s ga (bevel tip) | needle size 22s ga (bevel tip) |
| volume 50 μL | volume 10 μL | volume 25 μL | volume 100 μL |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Serie 800, abnehmbare Nadel
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 21496 | 04061832872834 |