The Metabolic pathways poster I received is 12 x 19 but the letters are so small as to make it unusable--at least for me. I love the concept and the clean look of the product but it is impractical and not useful at all. I ordered the larger sized poster as well but it has been on backorder for more than a year so I haven't received it yet. I'm very disappointed in the product.
Fortfahren mit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NA.28
UNSPSC Code:
41100000
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003
General description
This 22nd edition of the IUBMB-Nicholson Metabolic Pathways Chart features an enhanced ATP metabolism in mitochondria and chloroplasts. It illustrates the sequences of reactions involved in different aspects of metabolism and how they are integrated. The backbone of the map is the Glycolytic Pathway, followed by the TCA (Krebs) Cycle and the ElectronTransport System, which together produce most of the energy (ATP) required for all life. Most other aspects of metabolism are associated with this backbone and are differentiated by the use of color. Human metabolism is distinguished by the use of black arrows. Some 550 reactions are identified by their IUBMB Enzyme Commission (EC) numbers which are then indexed.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| publication info D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003 | publication info D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 1998 | publication info D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003 | publication info - |
Still not finding the right product?
Explore all of our products under Metabolic Pathways, 22nd Ed.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?