Fortfahren mit
1.09126
Shoulder bag
for use with MAS-100 Eco®
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
PackungsgrößeSKUVerfügbarkeitPreis
1 unit
Warenkorb auf Verfügbarkeit prüfen
€ 289,00
Über diesen Artikel
NACRES:
NA.74
UNSPSC Code:
41122409
€ 289,00
Warenkorb auf Verfügbarkeit prüfen
Weitere Informationen anfragen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
fabric bag
agency
non-compendial
product line
MAS-100®
feature
portable
packaging
pkg of 1 unit
manufacturer/tradename
MAS-100 Eco®
application(s)
air monitoring
food and beverages
compatibility
for use with MAS-100 Eco®
General description
Shoulder bag for MAS-100 Eco® is an additional accessory. The MAS-100 Eco® is an air sampler, designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The shoulder bag aids in easy and safe transport of the air sampler.
Application
Shoulder bag for MAS-100 Eco® is an additional accessory for Air Sampler that is used for microbial monitoring of ambient air in food and beverage industry. Microbial air sampling devices are extensively used in Food, Beverage as well as Environmental applications.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
1.092001.191331.17269
compatibility
for use with MAS-100 Eco®
compatibility
for use with MAS-100 NT® Ex
compatibility
for use with MAS-100 NT®
compatibility
for use with MAS-100 Eco®
packaging
pkg of 1 unit
packaging
pkg of 1 kit
packaging
-
packaging
-
material
fabric bag
material
plastic hardware
material
plastic hardware
material
plastic hardware
application(s)
air monitoring
food and beverages
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
feature
portable
feature
electronic
feature
electronic
feature
electronic
product line
MAS-100®
product line
-
product line
-
product line
MAS-100®
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Y000146904061833807989
A78608-100G04061833386538
A78608-500G04061833386545
109126000104022536780214