Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 each | Warenkorb auf Verfügbarkeit prüfen | € 205,00 |
Über diesen Artikel
NACRES:
NA.74
UNSPSC Code:
41103403
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
plastic hardware
agency
non-compendial
product line
MAS-100®
feature
electronic
manufacturer/tradename
MAS-100 Eco®
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Eco®
General description
The MAS-100 Eco® battery charger is an additional accessory for MAS-100 Eco® microbial air sampler. The battery charger for MAS-100 Eco® air sampler has an input voltage of 100 to 240 V/AC (47-63 Hz) and an output voltage of 5 V/DC (500 mA)
Application
The MAS-100 Eco® charger is an additional accessory for MAS-100 Eco® air sampler that is used for microbial monitoring of ambient air in controlled environments. Microbial air sampling devices are extensively used in Food, Beverage and Environmental applications.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| compatibility for use with MAS-100 Eco® | compatibility for use with 1060, 1062, 1063, & 1067 Air Samplers | compatibility for use with MAS-100 NT® Ex | compatibility for use with MAS-100 NT® |
| feature electronic | feature - | feature electronic | feature electronic |
| material plastic hardware | material - | material plastic hardware | material plastic hardware |
| agency non-compendial | agency - | agency non-compendial | agency non-compendial |
| manufacturer/tradename MAS-100 Eco® | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| product line MAS-100® | product line - | product line - | product line - |
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1172690001 | 04054839637896 |