Über diesen Artikel
NACRES:
NB.24
UNSPSC Code:
41104916
Sterility:
non-sterile
Packaging:
package of 1 ea
sterility
non-sterile
packaging
package of 1 ea
manufacturer/tradename
Sartorius VCA012
availability
available only in EU
L × O.D.
3 m × 4 mm
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename Sartorius VCA012 | manufacturer/tradename Sartorius VCA011 | manufacturer/tradename Sartorius | manufacturer/tradename Sartorius |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| packaging package of 1 ea | packaging pkg of 1 ea | packaging package of 100 ea | packaging package of 25 ea |
| availability available only in EU | availability available only in EU | availability - | availability - |
| L × O.D. 3 m × 4 mm | L × O.D. - | L × O.D. - | L × O.D. - |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?