Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
24111500
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
clear linear low-density polyethylene (LLDPE)
sterility
sterile
feature
label plain white
packaging
pkg of 600 ea (6 boxes of 100EA)
manufacturer/tradename
Whirl-Pak® B01603
bag W × L × thickness
15 cm × 38 cm × 0.102 mm
capacity
1000 mL (34 oz)
General description
Meets National Environmental Laboratory Accreditation Conference (NELAC) standard, June 2003.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Features and Benefits
- EPA approved
- Self-standing gusseted bag
- Sodium thiosulfate tablet in the bag for chlorine neutralization
- Built-in puncture proof closure
- Post-manufacturing sterilization for results you can trust.
Legal Information
Thio-Bag is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material clear linear low-density polyethylene (LLDPE) | material clear linear low-density polyethylene (LLDPE) | material - | material - |
| capacity 1000 mL (34 oz) | capacity 500 mL (17 oz) | capacity 300 mL , 300 mL capacity | capacity 100 mL , 100 mL capacity |
| packaging pkg of 600 ea (6 boxes of 100EA) | packaging pkg of 900 ea (9 boxes of 100EA) | packaging pkg of 100 ea | packaging pkg of 100 ea |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| bag W × L × thickness 15 cm × 38 cm × 0.102 mm | bag W × L × thickness - | bag W × L × thickness - | bag W × L × thickness - |
| manufacturer/tradename Whirl-Pak® B01603 | manufacturer/tradename Whirl-Pak® B01601 | manufacturer/tradename Whirl-Pak® B01254WA | manufacturer/tradename Whirl-Pak® B01402WA |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| WPB01603-600EA | 04065268794210 |