Merck
WPB01254WA
Whirl-Pak® Thio-Bag®, Waste Water Sampling Bag
300 mL capacity
Synonym(e):
Thio-Bag Water Sampler, Waste Water Sampling Bag
Größe auswählen
100 EA
€ 101,00
€ 101,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 101,00
About This Item
UNSPSC-Code:
24111500
NACRES:
NB.35
€ 101,00
Check Cart for Availability
Sterilität
sterile
Verpackung
pkg of 100 ea
Hersteller/Markenname
Nasco B01254WA
Parameter
-346-180 °F temp. range (-210 to 82 °C)
B × L × Dicke
11.5 cm × 23 cm × 0.064 mm
Kapazität
300 mL
300 mL capacity
Allgemeine Beschreibung
Meets National Environmental Laboratory Accreditation Conference (NELAC) standard, June 2003.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Leistungsmerkmale und Vorteile
  • EPA approved
  • Self-standing gusseted bag
  • Sodium thiosulfate tablet in the bag for chlorine neutralization
  • Built-in puncture proof closure
  • Post-manufacturing sterilization for results you can trust.
Rechtliche Hinweise
Thio-Bag is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)