Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 ea | Warenkorb auf Verfügbarkeit prüfen | € 162,00 |
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Drummond Scientific 3-000-752
Material:
aluminum (Anodized), blue
Packaging:
pack of 1 ea
€ 162,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
aluminum (Anodized), blue
packaging
pack of 1 ea
manufacturer/tradename
Drummond Scientific 3-000-752
capacity
1-100 μL
General description
Anodized aluminum body and screw-operated piston aspirates and dispenses up to 100 μL. Tapered silicone rubber neck holds all sizes of capillaries securely.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material aluminum (Anodized), blue | material plastic , silicone | material plastic , silicone | material glass |
| manufacturer/tradename Drummond Scientific 3-000-752 | manufacturer/tradename Drummond Scientific 4-000-200 | manufacturer/tradename Drummond Scientific 4-000-201 | manufacturer/tradename Drummond Scientific 5-000-1001-X10 |
| packaging pack of 1 ea | packaging - | packaging - | packaging pack of 100 ea |
| capacity 1-100 μL | capacity - | capacity - | capacity 10 μL |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Liyang Zhang et al.
Genome research, 28(1), 111-121 (2017-12-03)
The DNA-binding interfaces of the androgen (AR) and glucocorticoid (GR) receptors are virtually identical, yet these transcription factors share only about a third of their genomic binding sites and regulate similarly distinct sets of target genes. To address this paradox
Julio C Sanchez et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1863(8), 194566-194566 (2020-05-08)
The ATP-dependent BAF chromatin remodeling complex plays a critical role in gene regulation by modulating chromatin architecture, and is frequently mutated in cancer. Indeed, subunits of the BAF complex are found to be mutated in >20% of human tumors. The
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P4925-1EA | 04061833452660 |