Fortfahren mit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
Lineare Formel:
CH3COOC2H5
CAS-Nummer:
Molekulargewicht:
88.11
EC Number:
205-500-4
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
506104
grade
ACS reagent
vapor density
3 (20 °C, vs air)
vapor pressure
73 mmHg ( 20 °C)
assay
≥99.5%
autoignition temp.
801 °F
expl. lim.
2.2-11.5 %, 38 °F
impurities
Substances darkened by sulfuric acid, passes test, ≤0.0009 meq/g Titratable acid, ≤0.2% Water (H2O)
evapn. residue
≤0.003%
color
APHA: ≤10
refractive index
n20/D 1.3720 (lit.)
bp
76.5-77.5 °C (lit.)
mp
−84 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
SMILES string
CCOC(C)=O
InChI
1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
InChI key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Ethylacetat
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Lagerklasse
3 - Flammable liquids
wgk
WGK 1
flash_point_f
24.8 °F - closed cup
flash_point_c
-4 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
Junwei Liu et al.
Planta, 241(6), 1435-1451 (2015-02-27)
Strigolactone changes and cross talk with ABA unveil a picture of root-specific hormonal dynamics under stress. Strigolactones (SLs) are carotenoid-derived hormones influencing diverse aspects of development and communication with (micro)organisms, and proposed as mediators of environmental stimuli in resource allocation
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Tsukasa Horiyama et al.
PloS one, 9(9), e108642-e108642 (2014-09-27)
Escherichia coli produces the iron-chelating compound enterobactin to enable growth under iron-limiting conditions. After biosynthesis, enterobactin is released from the cell. However, the enterobactin export system is not fully understood. Previous studies have suggested that the outer membrane channel TolC


