Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 100 mg | Warenkorb auf Verfügbarkeit prüfen | € 449,00 |
Über diesen Artikel
Empirische Formel (Hill-System):
C9H21N3O · 2HCl
CAS-Nummer:
Molekulargewicht:
260.20
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3698131
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltengrade
analytical standard
Quality Segment
technique(s)
mass spectrometry (MS): suitable
format
neat
storage temp.
2-8°C
SMILES string
Cl.Cl.CC(=O)NCCCCNCCCN
InChI
1S/C9H21N3O.2ClH/c1-9(13)12-8-3-2-6-11-7-4-5-10;;/h11H,2-8,10H2,1H3,(H,12,13);2*1H
InChI key
BWGUIRARVGYOIR-UHFFFAOYSA-N
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| technique(s) mass spectrometry (MS): suitable | technique(s) mass spectrometry (MS): suitable | technique(s) mass spectrometry (MS): suitable | technique(s) mass spectrometry (MS): suitable |
| format neat | format - | format - | format - |
| Quality Level 200 | Quality Level - | Quality Level 200 | Quality Level - |
| grade analytical standard | grade - | grade - | grade - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Assay of N8-acetylspermidine deacetylase.
J Blankenship
Methods in molecular biology (Clifton, N.J.), 79, 77-80 (1998-02-17)
J Ekström et al.
Acta physiologica Scandinavica, 135(3), 255-261 (1989-03-01)
Polyamines are important for protein synthesis and tissue growth. In rat salivary glands, the activity of ornithine decarboxylase (ODC), the enzyme catalysing the formation of putrescine, and the content of putrescine, spermidine, spermine and N1-acetylspermidine were assayed after parasympathetic or
L Y Wing et al.
The Journal of pharmacology and experimental therapeutics, 266(1), 179-183 (1993-07-01)
Polyamines are polycations in cells and acetylation is the first step in their intracellular metabolism. We investigated the effects of the acetylated polyamines on arterial blood pressure and vascular reactivities in rats. Acetylspermine and acetylspermidine, administered at concentrations ranging from
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A3658-100MG | 04061832847467 |